missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gold powder, 99.>96%% (metals basis), Alfa Aesar™

Form: S.A. 0.45-0.70 m2/g

Manufacturer:  Thermo Fisher Scientific 4463603

 View more versions of this product

Catalog No. AA4463603



Specifications

Specifications

Gold
7440-57-5
19.32g/cm3 at 20°C
Au
burnish
[Au]
Odorless
23985
Sperical Powder, S.A. 0.45-0.70m
1g
Gold colored powder
>99.96% (Metals basis)
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
196.967
CHEBI:30050
Golden

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
burnish
CHEBI:30050
[Au]
Safety and Handling

Safety and Handling

EINECSNumber : 231-165-9

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents