missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold powder, -60 mesh, Premion™, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 1464106

 View more versions of this product

Catalog No. AA1464106



Specifications

Specifications

Gold powder
Premion™
(metals basis)
MFCD00003436
[Au]
196.967
CHEBI:30050
−60 Mesh
7440-57-5
99.99%
Au
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
23985
Powder
5g

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
CHEBI:30050
[Au]
Au
MFCD00003436
23985
gold
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents