missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold powder, -20 mesh, Premion™, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 1061203

 View more versions of this product

Catalog No. AA1061203



Specifications

Specifications

Gold powder
7440-57-5
Au
−20 Mesh
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
23985
Powder
Premion™
1g
(metals basis)
MFCD00003436
burnish
[Au]
196.967
CHEBI:30050
99.99%
Golden

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
burnish
CHEBI:30050
[Au]
Safety and Handling

Safety and Handling

RTECSNumber : MD5070000

TSCA : TSCA

Documents