missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gold Plasma Std. Solution, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 14392AD

 View more versions of this product

Catalog No. AA14392AD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Gold
Liquid
MFCD00003436
[Au]
196.967
CHEBI:30050
Characteristic
7440-57-5
Au
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
23985
Liquid
50mL

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
CHEBI:30050
[Au]
Au
MFCD00003436
23985
gold
Safety and Handling

Safety and Handling

H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.

DOTInformation : Hazard Class: 8, Packing Group: II

EINECSNumber : 231-595-7

Documents