missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gold Plasma Std. Solution, Alfa Aesar™, Specpure™
Manufacturer: Alfa Aesar 14392AP
Specifications
Gold | |
Liquid | |
MFCD00003436 | |
[Au] | |
196.967 | |
CHEBI:30050 | |
Characteristic | |
500mL |
7440-57-5 | |
Au | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
gold | |
23985 | |
Liquid | |
Approx. 1g/cm3 |
Chemical Identifiers
7440-57-5 | |
196.967 | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
CHEBI:30050 | |
[Au] |
Au | |
MFCD00003436 | |
23985 | |
gold |
Safety and Handling
H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.
DOTInformation : Hazard Class: 8, Packing Group: II
EINECSNumber : 231-595-7