Learn More
Gold, plasma standard solution, Specpure™ Au 1000μg/mL, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 013881AE
Description
Gold, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Gold plasma standard solution | |
Au 1000μg/mL | |
MFCD00003436 | |
Specpure™ | |
Liquid |
Au in 20% HCl | |
100 mL | |
UN1789 | |
Immiscible with water |
Chemical Identifiers
MFCD00003436 |
Safety and Handling
GHS H Statement
H314-H335
Causes severe skin burns and eye damage.
May cause respiratory irritation.
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROCHLORIC ACID
EINECSNumber : 231-595-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only