missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanorods, 9-15nm dia. x 55-65nm length, dispersion in water, 850nm Absorption, negatively charged

Manufacturer:  Alfa Aesar™ 46972AC

 View more versions of this product

Catalog No. AA46972AC



Specifications

Specifications

Gold nanorods
9 to 15nm dia. x 55 to 65nm length, dispersion in water, 850nm Absorption, negatively charged
Au
PCHJSUWPFVWCPO-UHFFFAOYSA-N
[Au]
196.967
CHEBI:30050
Colorless
25mL
7440-57-5
MFCD00003436
Odorless
gold
23985
Liquid
>60°C (decomposition)

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
CHEBI:30050
[Au]
Au
MFCD00003436
23985
gold
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents