missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanorods, 9-15nm dia. x 46-56nm length, dispersion in water, 800nm Absorption, negatively charged

Manufacturer:  Alfa Aesar™ 46819AC

 View more versions of this product

Catalog No. AA46819AC



Specifications

Specifications

Gold nanorods
9 to 15nm dia. x 46 to 56nm length, dispersion in water, 800nm Absorption, negatively charged
Au
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
Odorless
gold
23985
Liquid
>60°C (decomposition)
25mL
7440-57-5
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
[Au]
196.967
CHEBI:30050
Colorless

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
CHEBI:30050
[Au]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents