missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanorods, 19-25nm dia. x 50-60nm length, dispersion in water, 650nm Absorption, negatively charged

Manufacturer:  Alfa Aesar™ 46997AA

 View more versions of this product

Catalog No. AA46997AA



Specifications

Specifications

5mL
19 to 25nm dia. x 50 to 60nm length, dispersion in water, 650nm Absorption, negatively charged
Au
PCHJSUWPFVWCPO-UHFFFAOYSA-N
[Au]
196.967
CHEBI:30050
Colorless
Gold nanorods
7440-57-5
MFCD00003436
Odorless
gold
23985
Liquid
>60°C (decomposition)

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
CHEBI:30050
[Au]
Au
MFCD00003436
23985
gold
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents