missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 90nm, supplied in 0.1mg/mL sodium citrate with stabilizer, OD1, 564nm absorption

$96.76 - $298.95

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.967
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym burnish, c.i. pigment metal 3, ci pigment metal 3, colloidal, flake, gold, colloidal, leaf, magnesium purple, powder, shell
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ67184AC
View Documents
Alfa Aesar™
J67184AC
25mL Each for $96.76
 
AAJ67184AE
View Documents
Alfa Aesar™
J67184AE
100mL Each for $298.95
 
Description

Description

  • For adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols
  • Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications
Specifications

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
burnish, c.i. pigment metal 3, ci pigment metal 3, colloidal, flake, gold, colloidal, leaf, magnesium purple, powder, shell
CHEBI:30050
[Au]

Specifications

25mL
Au
burnish, c.i. pigment metal 3, ci pigment metal 3, colloidal, flake, gold, colloidal, leaf, magnesium purple, powder, shell
[Au]
196.967
CHEBI:30050
90nm
Pink to Red
7440-57-5
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
23985
Liquid
Light sensitive
Gold nanoparticles, supplied in 0.1mg/mL sodium citrate with stabilizer, OD: 1, 564nm absorption
Safety and Handling

Safety and Handling

TSCA : Yes

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold