missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 90nm, carboxy-functionalized, 3kDa PEGylated, OD50, 564nm absorption

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.967
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym burnish, c.i. pigment metal 3, ci pigment metal 3, colloidal, flake, gold, colloidal, leaf, magnesium purple, powder, shell
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ67300AMI
View Documents
Alfa Aesar™
J67300AMI
0.5mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ67300XF
View Documents
Alfa Aesar™
J67300XF
1mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
burnish, c.i. pigment metal 3, ci pigment metal 3, colloidal, flake, gold, colloidal, leaf, magnesium purple, powder, shell
CHEBI:30050
[Au]

Specifications

0.5mL
Au
burnish, c.i. pigment metal 3, ci pigment metal 3, colloidal, flake, gold, colloidal, leaf, magnesium purple, powder, shell
[Au]
196.967
CHEBI:30050
90nm
Pink to Red
7440-57-5
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
23985
Liquid
Light sensitive
Gold nanoparticles, carboxy-functionalized, 3kDa PEGylated, OD: 50, 564nm absorption
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS