Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 80nm, supplied in 0.1mM PBS, reactant-free, 99%, OD1, 553nm absorption

Chemical Identifiers

Chemical Name or Material Gold nanoparticles
CAS 7440-57-5
Name Note supplied in 0.1mM PBS, reactant-free, 99%, OD1, 553nm absorption
MDL Number MFCD00003436
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67068AC

 
alfa aesar™
J67068AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67068AE

 
alfa aesar™
J67068AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Gold nanoparticles
Name Note supplied in 0.1mM PBS, reactant-free, 99%, OD1, 553nm absorption
Physical Form Liquid
Assay 99%
MDL Number MFCD00003436
CAS 7440-57-5
Odor Odorless
Particle Size 80nm
Storage Note 1 2° to 8°C
Color Red to pink
Quantity 25mL
Safety and Handling

2° to 8°C