Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 80nm, carboxy-functionalized, 3kDa PEGylated, OD50, 553nm absorption

Chemical Identifiers

Chemical Name or Material Gold nanoparticles
Name Note carboxy-functionalized, 3kDa PEGylated, OD: 50, 553nm absorption
MDL Number MFCD00003436
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67108AMI

 
alfa aesar™
J67108AMI
0.5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67108XF

 
alfa aesar™
J67108XF
1mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Gold nanoparticles
Name Note carboxy-functionalized, 3kDa PEGylated, OD: 50, 553nm absorption
Physical Form Liquid
MDL Number MFCD00003436
Particle Size 80nm
Storage Note 1 2° to 8°C
Color Red to pink
Quantity 0.5mL
Safety and Handling

2° to 8°C