Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 70nm, supplied in 0.1mg/mL sodium citrate with stabilizer, OD1, 548nm absorption

Chemical Identifiers

Chemical Name or Material Gold nanoparticles
CAS 7440-57-5
Name Note supplied in 0.1mg/mL sodium citrate with stabilizer, OD: 1, 548nm absorption
MDL Number MFCD00003436
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67195AC

 
alfa aesar™
J67195AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67195AE

 
alfa aesar™
J67195AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Gold nanoparticles
CAS 7440-57-5
MDL Number MFCD00003436
Name Note supplied in 0.1mg/mL sodium citrate with stabilizer, OD: 1, 548nm absorption
Storage Note 1 2° to 8°C
Particle Size 70nm
Quantity 25mL
Color Red to pink
Physical Form Liquid

  • For use in adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols
  • Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications

Safety and Handling

2° to 8°C