Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 70nm, biotin-functionalized, PEGylated, OD50, supplied in purified water, 548 nm absorption

Chemical Identifiers

Chemical Name or Material Gold nanoparticles
Name Note biotin-functionalized, PEGylated, OD: 50, supplied in purified water, 548nm absorption
MDL Number MFCD00003436
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67374AMI

 
alfa aesar™
J67374AMI
0.5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67374XF

 
alfa aesar™
J67374XF
1mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Chemical Name or Material Gold nanoparticles
Name Note biotin-functionalized, PEGylated, OD: 50, supplied in purified water, 548nm absorption
MDL Number MFCD00003436
Particle Size 70nm
Storage Note 1 2° to 8°C
Color Red to pink
Quantity 0.5mL
Safety and Handling

2° to 8°C