Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 60nm, supplied in 0.1mM PBS, reactant-free, 99%, OD1, 540nm absorption

$125.51 - $125.51

Chemical Identifiers

Chemical Name or Material Gold nanoparticles
CAS 7440-57-5
Name Note supplied in 0.1mM PBS, reactant-free, 99%, OD1, 540nm absorption
MDL Number MFCD00003436
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67022AE

 
alfa aesar™
J67022AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67022AC

 
alfa aesar™
J67022AC
25mL Each for $125.51
Description & Specifications

Specifications

CAS 7440-57-5
Chemical Name or Material Gold nanoparticles
Physical Form Liquid
Assay 99%
MDL Number MFCD00003436
Name Note supplied in 0.1mM PBS, reactant-free, 99%, OD1, 540nm absorption
Particle Size 60nm
Storage Note 1 2° to 8°C
Color Red to pink
Quantity 100mL
Safety and Handling

2° to 8°C