Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 60nm, biotin-functionalized, PEGylated, OD50, supplied in purified water, 540nm absorption

Chemical Identifiers

Chemical Name or Material Gold nanoparticles
Name Note biotin-functionalized, PEGylated, OD: 50, supplied in purified water, 540nm absorption
MDL Number MFCD00003436
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67210AMI

 
alfa aesar™
J67210AMI
0.5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67210XF

 
alfa aesar™
J67210XF
1mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Gold nanoparticles
MDL Number MFCD00003436
Physical Form Liquid
Name Note biotin-functionalized, PEGylated, OD: 50, supplied in purified water, 540nm absorption
Quantity 0.5mL
Storage Note 1 2° to 8°C
Color Red to pink
Particle Size 60nm
Safety and Handling

2° to 8°C