Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 60nm, amine-functionalized, 3kDa PEGylated, OD50, 540nm absorption


Quantity 0.5mL

Manufacturer: alfa aesar™  J67365AMI

 View more versions of this product

Catalog No. AAJ67365AMI

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

MDL Number MFCD00003436
Name Note amine-functionalized, 3kDa PEGylated, OD: 50, 540nm absorption
Physical Form Liquid
Particle Size 60nm
Storage Note 1 2° to 8°C
Color Red to pink
Quantity 0.5mL
Chemical Name or Material Gold nanoparticles

Specifications

Chemical Name or Material Gold nanoparticles
Name Note amine-functionalized, 3kDa PEGylated, OD: 50, 540nm absorption
MDL Number MFCD00003436
Accessories

2° to 8°C