Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 5nm, supplied in 0.1mg/mL sodium citrate with stabilizer, OD1, 515-520nm absorption

Chemical Identifiers

Chemical Name or Material Gold nanoparticles
CAS 7440-57-5
Name Note supplied in 0.1mg/mL sodium citrate with stabilizer, OD: 1, 515 to 520nm absorption
MDL Number MFCD00003436
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67085AC

  SDS
alfa aesar™
J67085AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67085AE

  SDS
alfa aesar™
J67085AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Gold nanoparticles
CAS 7440-57-5
MDL Number MFCD00003436
Physical Form Liquid
Name Note supplied in 0.1mg/mL sodium citrate with stabilizer, OD: 1, 515 to 520nm absorption
Storage Note 1 2° to 8°C
Odor Odorless
Quantity 25mL
Color Red
TSCA TSCA
Particle Size 5nm

  • For use in adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols
  • Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications

Safety and Handling

2° to 8°C