missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gold nanoparticles, 5nm, supplied in 0.1mg/mL sodium citrate with stabilizer, OD1, 515-520nm absorption, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J67085AC
Description
For use in adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- For use in adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols
- Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications
Specifications
Red | |
supplied in 0.1mg/mL sodium citrate with stabilizer, OD: 1, 515 to 520nm absorption | |
Au | |
25 mL | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
gold | |
23985 | |
Odorless | |
Light sensitive |
Gold nanoparticles | |
7440-57-5 | |
MFCD00003436 | |
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
[Au] | |
196.97 | |
CHEBI:30050 | |
5nm | |
Liquid |
Chemical Identifiers
7440-57-5 | |
196.97 | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
23985 | |
gold |
Au | |
MFCD00003436 | |
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
CHEBI:30050 | |
[Au] |
Safety and Handling
TSCA : Yes
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only