missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gold nanoparticles, 50nm, supplied in 0.1 mg/ml sodium citrate with stabilizer OD1, 535nm absorption, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J67433AC
Description
For adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- For adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols
- Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications
Specifications
Pink to Red | |
supplied in 0.1mg/mL sodium citrate with stabilizer OD: 1, 535nm absorption | |
Au | |
25 mL | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
gold | |
23985 | |
50nm | |
Liquid |
Gold nanoparticles | |
7440-57-5 | |
MFCD00003436 | |
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
[Au] | |
196.97 | |
CHEBI:30050 | |
Light sensitive |
Chemical Identifiers
7440-57-5 | |
196.97 | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
23985 | |
gold |
Au | |
MFCD00003436 | |
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
CHEBI:30050 | |
[Au] |
Safety and Handling
TSCA : Yes
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only