missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 40nm, biotin-functionalized, PEGylated, OD50, supplied in purified water, 530nm absorption

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.967
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym burnish, c.i. pigment metal 3, ci pigment metal 3, colloidal, flake, gold, colloidal, leaf, magnesium purple, powder, shell
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ67026AMI
View Documents
Alfa Aesar™
J67026AMI
0.5mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ67026XF
View Documents
Alfa Aesar™
J67026XF
1mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
burnish, c.i. pigment metal 3, ci pigment metal 3, colloidal, flake, gold, colloidal, leaf, magnesium purple, powder, shell
CHEBI:30050
[Au]

Specifications

0.5mL
Au
burnish, c.i. pigment metal 3, ci pigment metal 3, colloidal, flake, gold, colloidal, leaf, magnesium purple, powder, shell
[Au]
196.967
CHEBI:30050
40nm
Pink to Red
7440-57-5
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
23985
Liquid
Light sensitive
Gold nanoparticles, biotin-functionalized, PEGylated, OD: 50, supplied in purified water, 530nm absorption
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS