Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 400nm, supplied in 0.1mM PBS, reactant-free, 99%

Chemical Identifiers

Chemical Name or Material Gold nanoparticles
CAS 7440-57-5
Name Note supplied in 0.1mM PBS, reactant-free
MDL Number MFCD00003436
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67324AC

  SDS
alfa aesar™
J67324AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67324AE

  SDS
alfa aesar™
J67324AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Gold nanoparticles
MDL Number MFCD00003436
Assay 99%
CAS 7440-57-5
TSCA TSCA
Physical Form Liquid
Particle Size 400nm
Storage Note 1 2° to 8°C
Odor Odorless
Quantity 25mL
Color Pink
Name Note supplied in 0.1mM PBS, reactant-free
Safety and Handling

2° to 8°C