Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 400nm, supplied in 0.1mg/mL sodium citrate with stabilizer

Chemical Identifiers

Chemical Name or Material Gold nanoparticles
CAS 7440-57-5
Name Note supplied in 0.1mg/mL sodium citrate with stabilizer
MDL Number MFCD00003436
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67213AC

 
alfa aesar™
J67213AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67213AE

 
alfa aesar™
J67213AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Gold nanoparticles
Physical Form Liquid
MDL Number MFCD00003436
CAS 7440-57-5
Storage Note 1 2° to 8°C
Color Pink
Name Note supplied in 0.1mg/mL sodium citrate with stabilizer
Particle Size 400nm
Quantity 25mL
Safety and Handling

2° to 8°C