Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 30nm, amine-functionalized, 3kDa PEGylated, OD50, 526nm absorption

Chemical Identifiers

Chemical Name or Material Gold nanoparticles
Name Note amine-functionalized, 3kDa PEGylated, OD: 50, 526nm absorption
MDL Number MFCD00003436
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67391AMI

 
alfa aesar™
J67391AMI
0.5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67391XF

 
alfa aesar™
J67391XF
1mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

MDL Number MFCD00003436
Physical Form Liquid
Name Note amine-functionalized, 3kDa PEGylated, OD: 50, 526nm absorption
Color Red
Quantity 0.5mL
Particle Size 30nm
Storage Note 1 2° to 8°C
Chemical Name or Material Gold nanoparticles
Safety and Handling

2° to 8°C