missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 30nm, amine-functionalized, 3kDa PEGylated, OD50, 526nm absorption

Manufacturer:  Alfa Aesar™ J67391XF

 View more versions of this product

Catalog No. AAJ67391XF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Gold nanoparticles
amine-functionalized, 3kDa PEGylated, OD: 50, 526nm absorption
Au
burnish, c.i. pigment metal 3, ci pigment metal 3, colloidal, flake, gold, colloidal, leaf, magnesium purple, powder, shell
[Au]
196.967
CHEBI:30050
30nm
Red
1mL
7440-57-5
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
23985
Liquid
Light sensitive

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
burnish, c.i. pigment metal 3, ci pigment metal 3, colloidal, flake, gold, colloidal, leaf, magnesium purple, powder, shell
CHEBI:30050
[Au]
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents