Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 300nm, supplied in 0.1mM PBS, reactant-free, 99%


Quantity 100mL

Manufacturer: alfa aesar™  J67137AE

 View more versions of this product

Catalog No. AAJ67137AE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Gold nanoparticles
Assay 99%
CAS 7440-57-5
Name Note supplied in 0.1mM PBS, reactant-free
Physical Form Liquid
MDL Number MFCD00003436
Quantity 100mL
Particle Size 300nm
Storage Note 1 2° to 8°C
Color Pink

Specifications

Chemical Name or Material Gold nanoparticles
CAS 7440-57-5
Name Note supplied in 0.1mM PBS, reactant-free
MDL Number MFCD00003436
Accessories

2° to 8°C