Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 300nm, supplied in 0.1mM PBS ,95%

Chemical Identifiers

Chemical Name or Material Gold nanoparticles
CAS 7440-57-5
Name Note supplied in 0.1mM PBS
MDL Number MFCD00003436
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67400AC

 
alfa aesar™
J67400AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67400AE

 
alfa aesar™
J67400AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Gold nanoparticles
MDL Number MFCD00003436
CAS 7440-57-5
Name Note supplied in 0.1mM PBS
Physical Form Liquid
Storage Note 1 2° to 8°C
Assay 95%
Particle Size 300nm
Color Bright red to opaque pink
Quantity 25mL

  • For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM

Safety and Handling

2° to 8°C