missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gold nanoparticles, 20nm, supplied in 0.1 mg/ml sodium citrate with stabilizer, OD1, 524nm absorption, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals J67269AC

 View more versions of this product

Catalog No. AAJ67269AC



Description

Description

Colloidal gold is an electron-dense, non-fading marker useful as a probe in electron microscopy, light microscopy and immunoblotting. It requires no additional processing for detection, but in some applications the signal can be enhanced by reaction with silver. It can be complexed with biomolecules by strong, non-covalent interactions. Particles less than 15 nm are most useful in transmission electron microscopy and for applications where access to the probe may be hindered by larger particles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Colloidal gold is an electron-dense, non-fading marker useful as a probe in electron microscopy, light microscopy and immunoblotting. It requires no additional processing for detection, but in some applications the signal can be enhanced by reaction with silver. It can be complexed with biomolecules by strong, non-covalent interactions. Particles less than 15 nm are most useful in transmission electron microscopy and for applications where access to the probe may be hindered by larger particles.

Solubility
Fully miscible in water. It is soluble in aqua regia, nitric acid and hydrochloric acid.

Notes
Particles per ml: 6.54E+11
Specifications

Specifications

Pink to Red
supplied in 0.1mg/mL sodium citrate with stabilizer, OD: 1, 524nm absorption
Au
25 mL
Fully miscible in water. It is soluble in aqua regia,nitric acid and hydrochloric acid.
[Au]
196.97
CHEBI:30050
Light sensitive
Gold nanoparticles
7440-57-5
MFCD00003436
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
23985
20nm
Liquid

Chemical Identifiers

7440-57-5
196.97
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
CHEBI:30050
[Au]
Safety and Handling

Safety and Handling

TSCA : Yes

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only