Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 20nm, carboxy-functionalized, 3kDa PEGylated, OD50, 524nm absorption


Quantity 1mL

Manufacturer: alfa aesar™  J67386XF

 View more versions of this product

Catalog No. AAJ67386XF

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Gold nanoparticles
MDL Number MFCD00003436
Storage Note 1 2° to 8°C
Name Note carboxy-functionalized, 3kDa PEGylated, OD: 50, 524nm absorption
Color Red
Particle Size 20nm
Quantity 1mL
Physical Form Liquid

Specifications

Chemical Name or Material Gold nanoparticles
Name Note carboxy-functionalized, 3kDa PEGylated, OD: 50, 524nm absorption
MDL Number MFCD00003436
Accessories

2° to 8°C