missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 20nm, amine-functionalized, 3kDa PEGylated, OD50, 524nm absorption

Manufacturer:  Alfa Aesar™ J67465XF

 View more versions of this product

Catalog No. AAJ67465XF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Gold nanoparticles
amine-functionalized, 3kDa PEGylated, OD: 50, 524nm absorption
Au
burnish
[Au]
196.967
CHEBI:30050
20nm
1mL
7440-57-5
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
23985
Liquid
Red

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
burnish
CHEBI:30050
[Au]
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

Documents