Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 200nm, supplied in 0.1mM PBS, reactant-free, 99%

Chemical Identifiers

Chemical Name or Material Gold nanoparticles
CAS 7440-57-5
Name Note supplied in 0.1mM PBS, reactant-free
MDL Number MFCD00003436
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67446AC

 
alfa aesar™
J67446AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67446AE

 
alfa aesar™
J67446AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Gold nanoparticles
CAS 7440-57-5
Name Note supplied in 0.1mM PBS, reactant-free
Physical Form Liquid
Assay 99%
MDL Number MFCD00003436
Particle Size 200nm
Storage Note 1 2° to 8°C
Color Dark brown to pink
Quantity 25mL
Safety and Handling

2° to 8°C