Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 200nm, supplied in 0.1mg/mL sodium citrate with stabilizer

Chemical Identifiers

Chemical Name or Material Gold nanoparticles
CAS 7440-57-5
Name Note supplied in 0.1mg/mL sodium citrate with stabilizer
MDL Number MFCD00003436
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67327AC

 
alfa aesar™
J67327AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67327AE

 
alfa aesar™
J67327AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Gold nanoparticles
CAS 7440-57-5
Name Note supplied in 0.1mg/mL sodium citrate with stabilizer
Physical Form Liquid
Storage Note 1 2° to 8°C
Color Dark brown to pink
Quantity 25mL
Particle Size 200nm
MDL Number MFCD00003436

  • For adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols
  • Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications

Safety and Handling

2° to 8°C