Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 15nm, supplied in 0.1mg/mL sodium citrate with stabilizer, OD1, 520nm absorption


Quantity 100mL

Manufacturer: alfa aesar™  J67307AE

 View more versions of this product

Catalog No. AAJ67307AE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Gold nanoparticles
CAS 7440-57-5
Name Note supplied in 0.1mg/mL sodium citrate with stabilizer, OD: 1, 520nm absorption
Physical Form Liquid
MDL Number MFCD00003436
TSCA TSCA
Particle Size 15nm
Storage Note 1 2° to 8°C
Color Pink to red
Quantity 100mL

Specifications

Chemical Name or Material Gold nanoparticles
CAS 7440-57-5
Name Note supplied in 0.1mg/mL sodium citrate with stabilizer, OD: 1, 520nm absorption
MDL Number MFCD00003436

  • For use in adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols
  • Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications

Accessories

2° to 8°C

SDS & Safety Documents