Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 15nm, amine-functionalized, 3kDa PEGylated, 0D50, 520nm absorption


Quantity 1mL

Manufacturer: alfa aesar™  J67326XF

 View more versions of this product

Catalog No. AAJ67326XF

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

MDL Number MFCD00003436
Physical Form Liquid
Chemical Name or Material Gold nanoparticles
Name Note amine-functionalized, 3kDa PEGylated, 0D50, 520nm absorption
Particle Size 15nm
Color Red
Quantity 1mL
Storage Note 1 2° to 8°C

Specifications

Chemical Name or Material Gold nanoparticles
Name Note amine-functionalized, 3kDa PEGylated, 0D50, 520nm absorption
MDL Number MFCD00003436
Accessories

2° to 8°C