Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 150nm, supplied in 0.1mg/mL sodium citrate with stabilizer


Quantity 100mL

Manufacturer: alfa aesar™  J67342AE

 View more versions of this product

Catalog No. AAJ67342AE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Gold nanoparticles
MDL Number MFCD00003436
CAS 7440-57-5
Name Note supplied in 0.1mg/mL sodium citrate with stabilizer
Physical Form Liquid
Particle Size 150nm
Storage Note 1 2° to 8°C
Color Dark brown to pink
Quantity 100mL

Specifications

Chemical Name or Material Gold nanoparticles
CAS 7440-57-5
Name Note supplied in 0.1mg/mL sodium citrate with stabilizer
MDL Number MFCD00003436
Accessories

2° to 8°C