Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 10nm, supplied in 0.1mg/mL sodium citrate with stabilizer, OD1, 520nm absorption


Quantity 25mL

Manufacturer: alfa aesar™  J67188AC

 View more versions of this product

Catalog No. AAJ67188AC

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Gold nanoparticles
CAS 7440-57-5
MDL Number MFCD00003436
Physical Form Liquid
Storage Note 1 2° to 8°C
Particle Size 10nm
Odor Odorless
TSCA TSCA
Quantity 25mL
Color Pink to red
Name Note supplied in 0.1mg/mL sodium citrate with stabilizer, OD: 1, 520nm absorption

Specifications

Chemical Name or Material Gold nanoparticles
CAS 7440-57-5
Name Note supplied in 0.1mg/mL sodium citrate with stabilizer, OD: 1, 520nm absorption
MDL Number MFCD00003436

  • For use in adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols
  • Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications

Accessories

2° to 8°C

SDS & Safety Documents