Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 10nm, biotin-functionalized, PEGylated, OD50, supplied in purified water, 520 nm absorption

Chemical Identifiers

Chemical Name or Material Gold nanoparticles
Name Note biotin-functionalized, PEGylated, OD: 50, supplied in purified water, 520nm absorption
MDL Number MFCD00003436
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67408AMI

 
alfa aesar™
J67408AMI
0.5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67408XF

 
alfa aesar™
J67408XF
1mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

MDL Number MFCD00003436
Physical Form Liquid
Storage Note 1 2° to 8°C
Color Red
Name Note biotin-functionalized, PEGylated, OD: 50, supplied in purified water, 520nm absorption
Quantity 0.5mL
Particle Size 10nm
Chemical Name or Material Gold nanoparticles
Safety and Handling

2° to 8°C