Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 10nm, amine-functionalized, 3kDa PEGylated, D50, 520nm absorption


Quantity 0.5mL

Manufacturer: alfa aesar™  J67310AMI

 View more versions of this product

Catalog No. AAJ67310AMI

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Gold nanoparticles
Physical Form Liquid
Name Note amine-functionalized, 3kDa PEGylated, D50, 520nm absorption
Color Red
Quantity 0.5mL
Storage Note 1 2° to 8°C
Particle Size 10nm
MDL Number MFCD00003436

Specifications

Chemical Name or Material Gold nanoparticles
Name Note amine-functionalized, 3kDa PEGylated, D50, 520nm absorption
MDL Number MFCD00003436
Accessories

2° to 8°C