Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 100nm, supplied in 0.1mM PBS, 95%, OD1, 572nm absorption


Quantity 100mL

Manufacturer: alfa aesar™  J67348AE

 View more versions of this product

Catalog No. AAJ67348AE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Gold nanoparticles
MDL Number MFCD00003436
CAS 7440-57-5
Physical Form Liquid
Particle Size 100nm
Name Note supplied in 0.1mM PBS, 95%, OD: 1, 572nm absorption
Quantity 100mL
Storage Note 1 2° to 8°C
Color Red to pink
Assay 95%

Specifications

Chemical Name or Material Gold nanoparticles
CAS 7440-57-5
Name Note supplied in 0.1mM PBS, 95%, OD: 1, 572nm absorption
MDL Number MFCD00003436

  • For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM

Accessories

2° to 8°C

SDS & Safety Documents