missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 100nm, carboxy-functionalized, 3kDa PEGylated, OD50, 572nm absorption

Manufacturer:  Alfa Aesar™ J67153XF

 View more versions of this product

Catalog No. AAJ67153XF

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Gold nanoparticles
carboxy-functionalized, 3kDa PEGylated, OD: 50, 572nm absorption
MFCD00003436
100nm
1mL
Liquid
Odorless
Red to pink

Chemical Identifiers

MFCD00003436
Gold nanoparticles, carboxy-functionalized, 3kDa PEGylated, OD: 50, 572nm absorption

Safety and Handling

SDS & Safety Documents

storageNote1 : 2° to 8°C