Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold nanoparticles, 100nm, amine-functionalized, 3kDa PEGylated, OD50, 572nm absorption


Quantity 1mL

Manufacturer: alfa aesar™  J67484XF

 View more versions of this product

Catalog No. AAJ67484XF

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Gold nanoparticles
Physical Form Liquid
Particle Size 100nm
Name Note amine-functionalized, 3kDa PEGylated, OD: 50, 572nm absorption
Storage Note 1 2° to 8°C
Quantity 1mL
Color Red to pink
MDL Number MFCD00003436

Specifications

Chemical Name or Material Gold nanoparticles
Name Note amine-functionalized, 3kDa PEGylated, OD: 50, 572nm absorption
MDL Number MFCD00003436
Accessories

2° to 8°C