missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gold(III) chloride, Au 64.4% min, Alfa Aesar

Manufacturer:  Alfa Aesar 1216301

 View more versions of this product

Catalog No. AA1216301



Specifications

Specifications

Gold(III) chloride
AuCl3
Red crystalline
Crystalline
Red
13453-07-1
MFCD00014172
303.33
Odorless
0.25g

Chemical Identifiers

13453-07-1
AuCl3
MFCD00014172
Safety and Handling

Safety and Handling

H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.

DOTInformation : DOT Class: 8, Packing Group: III

EINECSNumber : 236-623-1

storageNote1 : Hygroscopic

Documents