Please login to your online account to display your discounted pricing

Gold(III) chloride, Au 64.4%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Gold(III) chloride
CAS 13453-07-1
Molecular Formula AuCl3
Formula Weight 303.33
MDL Number MFCD00014172
UN Number UN3260
Merck Index 14,4521
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1216306

  SDS
alfa aesar
1216306
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 303.33
Melting Point 180°C (subl.)
Quantity 5g
Chemical Name or Material Gold(III) chloride
TSCA TSCA
Odor Odorless
Physical Form Crystalline
Sensitivity Hygroscopic
Assay Percent Notes Au ≥64.4%
Molecular Formula AuCl3
MDL Number MFCD00014172
CAS 13453-07-1
UN Number UN3260
Merck Index 14,4521
Color Red
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 3.90
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
RTECS Number MD5420000
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.