Please login to your online account to display your discounted pricing

Gold(III) chloride, 99%, ACROS Organics™


Quantity 500mg
Packaging Glass bottle

Manufacturer: fisher scientific acros  318645000

 View more versions of this product

Catalog No. AC318645000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Gold(III) chloride
Name Note 99%
CAS 13453-07-1
Health Hazard 1 HYGROSCOPIC
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECS Number MD5420000
EINECS Number 236-623-1
TSCA TSCA
Physical Form Crystalline Powder, Crystals or Chunks
Assay ≥98.5% (ex Au)
Assay Percent Range 99%
Molecular Formula AuCl3
Linear Formula AuCl3
Formula Weight 303.32
MDL Number MFCD00014172
Merck Index 15, 4556
Packaging Glass bottle
Color Light yellow to beige to light brown
Quantity 500mg

Specifications

Chemical Name or Material Gold(III) chloride
Name Note 99%
CAS 13453-07-1
Assay ≥98.5% (ex Au)
Assay Percent Range 99%
Molecular Formula AuCl3
Linear Formula AuCl3
Formula Weight 303.32
MDL Number MFCD00014172
Accessories

HYGROSCOPIC

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents