missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gold(III) chloride, 99.99% (metals basis), Au 64.4% min, Alfa Aesar™, Premion™

Manufacturer:  Alfa Aesar 4336006

 View more versions of this product

Catalog No. AA4336006



Specifications

Specifications

Gold(III) Chloride
99.99% (Metal basis)
MFCD00014172
Red crystalline
Crystalline
Odorless
160°C
13453-07-1
AuCl3
3260
303.33
3.90g/cm3
Red
5g

Chemical Identifiers

13453-07-1
AuCl3
MFCD00014172
Safety and Handling

Safety and Handling

H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.

DOTInformation : DOT Class: 8, Packing Group: III

EINECSNumber : 236-623-1

Documents