missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold(III) bromide, Premion™, 99.99% (metals basis), Au 44.6% min

Manufacturer:  Alfa Aesar™ 1216203

 View more versions of this product

Catalog No. AA1216203



Specifications

Specifications

Gold(III) bromide
(metals basis)
UN3260
Granular
Premion™
~160°C (decomposition)
10294-28-7
MFCD00014171
436.69
99.99%
Au 44.6% min.
1g

Chemical Identifiers

10294-28-7
MFCD00014171
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

Documents