Learn More
Gold(III) bromide, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1951406
Description
Gold(III) bromide is used as a catalyst in a variety of reactions. It catalyzes the reaction between an enynal unit and carbonyl compounds to form a six-membered cyclic compound. Another catalytic use of gold tribromide is in the nucleophilic substitution reaction of propargylic alcohols, with the gold complex acting as an alcohol-activating agent to facilitate the substitution. It can be used as a testing reagent for the presence of ketamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGold(III) bromide is used as a catalyst in a variety of reactions. It catalyzes the reaction between an enynal unit and carbonyl compounds to form a six-membered cyclic compound. Another catalytic use of gold tribromide is in the nucleophilic substitution reaction of propargylic alcohols, with the gold complex acting as an alcohol-activating agent to facilitate the substitution. It can be used as a testing reagent for the presence of ketamine.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
Specifications
Gold(III) bromide | |
10294-28-7 | |
AuBr3H3 | |
UN3260 | |
KJCSJLXVPHOFJC-UHFFFAOYSA-N | |
gold(III) bromide | |
436.69 | |
Hygroscopic |
∼160°C (decomposition) | |
5 g | |
MFCD00014171 | |
Slightly soluble in water. | |
Br.Br.Br.[Au] | |
439.70 | |
99% |
Chemical Identifiers
10294-28-7 | |
439.70 | |
KJCSJLXVPHOFJC-UHFFFAOYSA-N | |
Br.Br.Br.[Au] |
AuBr3H3 | |
MFCD00014171 | |
gold(III) bromide |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 233-654-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only