missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gold(III) acetate, 99.9% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03974206
Description
Used in solar energy technologies. Used in petrochemical cracking and automotive catalysts, water treatment, plating, textiles, research and in optic, laser, crystal and glass applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Gold(III) acetate | |
15804-32-7 | |
5 g | |
C6H9AuO6 | |
gold 3+ triacetate, gold iii acetate, gold acetate, acmc-1c3i2, triacetic acid gold iii salt, acetic acid, gold 3+ salt 3:1 | |
OTCKNHQTLOBDDD-UHFFFAOYSA-K | |
gold(3+);triacetate | |
15952561 | |
Brown powder |
Brown | |
Powder | |
99.9% (Metal basis) | |
MFCD00210614 | |
Slightly soluble in water. | |
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Au+3] | |
374.099 | |
374.1 |
Chemical Identifiers
15804-32-7 | |
374.099 | |
OTCKNHQTLOBDDD-UHFFFAOYSA-K | |
15952561 | |
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Au+3] |
C6H9AuO6 | |
MFCD00210614 | |
gold 3+ triacetate, gold iii acetate, gold acetate, acmc-1c3i2, triacetic acid gold iii salt, acetic acid, gold 3+ salt 3:1 | |
gold(3+);triacetate |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only