Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold(I) sodium cyanide, 99.9% (metals basis), Au 72% min

$327.86 - $327.86

Chemical Identifiers

Chemical Name or Material Gold(I) sodium cyanide
CAS 15280-09-8
Linear Formula NaAu(CN)2
Formula Weight 271.99
MDL Number MFCD00050423
UN Number UN1588
Merck Index 14,8610
Synonym Sodium gold cyanide; Sodium dicyanoaurate(I)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1263703

 
alfa aesar™
1263703
1g Each for $327.86

AA1263701

 
alfa aesar™
1263701
0.25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1263706

 
alfa aesar™
1263706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 271.99
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Gold(I) sodium cyanide
TSCA TSCA
Odor Characteristic
Physical Form Powder
Assay 99.9%
Assay Percent Notes (metals basis)
Linear Formula NaAu(CN)2
MDL Number MFCD00050423
CAS 15280-09-8
UN Number UN1588
Merck Index 14,8610
Synonym Sodium gold cyanide; Sodium dicyanoaurate(I)
Quantity 1g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Concentration Au 72% min.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CYANIDES, INORGANIC, SOLID, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.